Order Entry
Export
ContactUsLinkComponent
15-Bromo-1,1,1,2,2,3,3,4,4-nonafluoropentadecane 95%
Irritant
15-Bromo-1,1,1,2,2,3,3,4,4-nonafluoropentadecane 95%
Catalog # APOSPC53053-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
15-Bromo-1,1,1,2,2,3,3,4,4-nonafluoropentadecane 95%
Catalog # APOSPC53053-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :453.225 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:213207-95-5
Formula:C₁₅H₂₂BrF₉
MDL Number:MFCD28963747