2,5-Difluoro-L-phenylalanine 97%
Catalog # APOSPC530006-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:201.17 g/mol
Storage Temperature:Ambient
CAS Number:31105-92-7
Formula:C₉H₉F₂NO₂
MDL Number:MFCD07372000