
2,2-Difluoro-1-(4-fluorophenyl)ethanol
Catalog # APOSPC52221-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:176.136 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:2546-44-3
Formula:C₈H₇F₃O
MDL Number:MFCD16618132