
5,6-Difluoro-1H-1,3-benzodiazole-2-carbaldehyde
Catalog # APOSPC52218-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:182.127 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1263379-05-0
Formula:C₈H₄F₂N₂O
MDL Number:MFCD18432482