Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-4,5-difluorobenzal fluoride
3-Bromo-4,5-difluorobenzal fluoride
3-Bromo-4,5-difluorobenzal fluoride
Irritant
3-Bromo-4,5-difluorobenzal fluoride
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Bromo-4,5-difluorobenzal fluoride
Supplier:
Apollo Scientific
About this item
CAS Number:
1782865-38-6
:
Formula:
C₇H₃BrF₄
MDL Number:
MFCD28737888
:
242.996 g/mol
:
GHS07
Storage Temperature:
Ambient