Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 2,2-difluoro-3-methyl-butanoate
Ethyl 2,2-difluoro-3-methyl-butanoate
Ethyl 2,2-difluoro-3-methyl-butanoate
Ethyl 2,2-difluoro-3-methyl-butanoate
APOSPC52101-1G
:
Apollo Scientific
:
Ethyl 2,2-difluoro-3-methyl-butanoate
APOSPC52101-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
166.166 g/mol
:
Refrigerator
:
1993
:
3,III
:
GHS02
:
165544-22-9
:
C₇H₁₂F₂O₂
:
MFCD11007689