tert-Butyl N-[3,3-difluoro-1-(hydroxymethyl)cyclobutyl]carbamate
Catalog # APOSPC520867-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:237.244 g/mol
Storage Temperature:Ambient
CAS Number:1232365-42-2
Formula:C₁₀H₁₇F₂NO₃
MDL Number:MFCD22566207