Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(2-Bromophenyl)-2,2-difluoro-ethanol
2-(2-Bromophenyl)-2,2-difluoro-ethanol 94%
2-(2-Bromophenyl)-2,2-difluoro-ethanol 94%
APOSPC520829-250MG
:
Apollo Scientific
:
2-(2-Bromophenyl)-2,2-difluoro-ethanol 94%
APOSPC520829-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
237.041 g/mol
:
Ambient
:
C₈H₇BrF₂O
:
MFCD28038787