Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(4-Bromophenyl)-2,2-difluoro-ethanol
2-(4-Bromophenyl)-2,2-difluoro-ethanol 97%
2-(4-Bromophenyl)-2,2-difluoro-ethanol 97%
2-(4-Bromophenyl)-2,2-difluoro-ethanol 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2-(4-Bromophenyl)-2,2-difluoro-ethanol 97%
Supplier:
Apollo Scientific
About this item
:
Formula:
C₈H₇BrF₂O
MDL Number:
MFCD28038792
:
237.041 g/mol
Storage Temperature:
Ambient