2-[2-(Difluoromethyl)-3,4-Difluoro-phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Catalog # APOSPC520321-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:290.062 g/mol
Storage Temperature:Ambient
Formula:C₁₃H₁₅BF₄O₂
MDL Number:MFCD30496636