Order Entry
Export
ContactUsLinkComponent
2-[5-(Difluoromethyl)-4-fluoro-2-methyl-phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-[5-(Difluoromethyl)-4-fluoro-2-methyl-phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Catalog # APOSPC520320-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[5-(Difluoromethyl)-4-fluoro-2-methyl-phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Catalog # APOSPC520320-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :286.098 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₈BF₃O₂
MDL Number:MFCD30496635