Order Entry
Export
ContactUsLinkComponent
1,3-Dibenzyloxy-5-(difluoromethyl)benzene
1,3-Dibenzyloxy-5-(difluoromethyl)benzene
Catalog # APOSPC520305-1G
Supplier:  Apollo Scientific
CAS Number:  
1,3-Dibenzyloxy-5-(difluoromethyl)benzene
Catalog # APOSPC520305-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :340.363 g/mol
Storage Temperature:Ambient
CAS Number:2149601-37-4
Formula:C₂₁H₁₈F₂O₂
MDL Number:MFCD30496623