4-(Difluoromethyl)-2,6-Difluoro-benzene-1,3-diol
Catalog # APOSPC520294-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.099 g/mol
Storage Temperature:Ambient
CAS Number:2092183-23-6
Formula:C₇H₄F₄O₂
MDL Number:MFCD30496612