2,3-Dibromo-5-(difluoromethyl)thiophene
Catalog # APOSPC520249-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:291.939 g/mol
Storage Temperature:Ambient
Formula:C₅H₂Br₂F₂S
MDL Number:MFCD29767728