Order Entry
Export
ContactUsLinkComponent
 
2-(Difluoromethyl)-7-methyl-imidazo[1,2-a]pyridine
Catalog # APOSPC520147-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Difluoromethyl)-7-methyl-imidazo[1,2-a]pyridine
Catalog # APOSPC520147-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :182.17 g/mol
Storage Temperature:Ambient
Formula:C₉H₈F₂N₂
MDL Number:MFCD29767628