5-(Difluoromethyl)pyrimidine-2,4-diamine
Catalog # APOSPC520064-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:160.125 g/mol
Storage Temperature:Ambient
Formula:C₅H₆F₂N₄
MDL Number:MFCD29767547