4,6-Dichloro-5-(difluoromethyl)-2-pyrrolidin-1-yl-pyrimidine
Catalog # APOSPC520055-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:268.091 g/mol
Storage Temperature:Ambient
CAS Number:2091581-62-1
Formula:C₉H₉Cl₂F₂N₃
MDL Number:MFCD29767538