4,6-Dichloro-5-(difluoromethyl)pyrimidin-2-amine
Catalog # APOSPC520048-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:214 g/mol
Storage Temperature:Ambient
CAS Number:2091217-55-7
Formula:C₅H₃Cl₂F₂N₃
MDL Number:MFCD29767531