4,6-Dichloro-5-(difluoromethyl)-2-methyl-pyrimidine
Catalog # APOSPC520045-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:213.012 g/mol
Storage Temperature:Ambient
CAS Number:2091563-68-5
Formula:C₆H₄Cl₂F₂N₂
MDL Number:MFCD29767528