3-Bromo-6-(difluoromethyl)imidazo[1,2-a]pyridine
Catalog # APOSPC520027-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:247.04 g/mol
Storage Temperature:Ambient
CAS Number:2090187-13-4
Formula:C₈H₅BrF₂N₂
MDL Number:MFCD29767511