Order Entry
Export
ContactUsLinkComponent
 
Irritant
3,4-Diaminophenylsulphur pentafluoride 97%
Catalog # APOSPC5149-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3,4-Diaminophenylsulphur pentafluoride 97%
Catalog # APOSPC5149-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :234.19 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:663179-59-7
Formula:C₆H₇F₅N₂S
MDL Number:MFCD11501040