Order Entry
Export
ContactUsLinkComponent
tert-Butyl 4-amino-3,3-difluoropyrrolidine-1-carboxylate 97%
tert-Butyl 4-amino-3,3-difluoropyrrolidine-1-carboxylate 97%
Catalog # APOSPC51388-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 4-amino-3,3-difluoropyrrolidine-1-carboxylate 97%
Catalog # APOSPC51388-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :222.232 g/mol
Storage Temperature:Refrigerator
CAS Number:1408074-83-8
Formula:C₉H₁₆F₂N₂O₂
MDL Number:MFCD23105898