2-[4-(Trifluoromethoxy)phenyl]-1,3,8-triazaspiro[4.5]dec-1-en-4-one 97%
Catalog # APOSPC51131-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:313.275 g/mol
Storage Temperature:Ambient
CAS Number:1253924-71-8
Formula:C₁₄H₁₄F₃N₃O₂
MDL Number:MFCD22375530