Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-1,1-difluorobut-1-ene
4-Chloro-1,1-difluorobut-1-ene 97%
4-Chloro-1,1-difluorobut-1-ene 97%
APOSPC51109-1G
:
Apollo Scientific
:
4-Chloro-1,1-difluorobut-1-ene 97%
APOSPC51109-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
126.532 g/mol
:
73…74 °C
:
Ambient
:
1993
:
3,II
:
GHS02
:
GHS07
:
1346521-43-4
:
C₄H₅ClF₂
:
MFCD20489353