Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Difluoro-5-iodonitrobenzene
2,4-Difluoro-5-iodonitrobenzene
2,4-Difluoro-5-iodonitrobenzene
2,4-Difluoro-5-iodonitrobenzene
Supplier:
Apollo Scientific
Certificates
Expand image
2,4-Difluoro-5-iodonitrobenzene
Supplier:
Apollo Scientific
About this item
CAS Number:
148388-74-3
:
Formula:
C₆H₂F₂INO₂
MDL Number:
MFCD20322039
:
284.987 g/mol
Boiling Pt:
65…67 °C (0.1 mm Hg)
:
GHS07
Storage Temperature:
Ambient