Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Dibromo-1H-tridecafluorooct-1-ene
1,2-Dibromo-1H-tridecafluorooct-1-ene
1,2-Dibromo-1H-tridecafluorooct-1-ene
1,2-Dibromo-1H-tridecafluorooct-1-ene
Supplier:
Apollo Scientific
Certificates
Expand image
1,2-Dibromo-1H-tridecafluorooct-1-ene
Supplier:
Apollo Scientific
About this item
CAS Number:
59665-28-0
:
Formula:
C₈HBr₂F₁₃
MDL Number:
MFCD18074336
:
503.881 g/mol
Storage Temperature:
Ambient