
4-Chloro-5,7-difluoroquinoline
Catalog # APOSPC51041-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:199.585 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:874831-46-6
Formula:C₉H₄ClF₂N
MDL Number:MFCD06808522