Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
8,8-Difluoro-1,4-dioxaspiro[4.5]decane
8,8-Difluoro-1,4-dioxaspiro[4.5]decane
8,8-Difluoro-1,4-dioxaspiro[4.5]decane
Irritant
8,8-Difluoro-1,4-dioxaspiro[4.5]decane
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
8,8-Difluoro-1,4-dioxaspiro[4.5]decane
Supplier:
Apollo Scientific
About this item
CAS Number:
176251-49-3
:
Formula:
C₈H₁₂F₂O₂
MDL Number:
MFCD11847478
:
178.176 g/mol
Melting Pt:
40…44 °C
:
GHS07
Storage Temperature:
Refrigerator