Ethyl 2,2-Difluoro-2-(4-methoxyphenyl)acetate
Catalog # APOSPC510342-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:230.208 g/mol
Storage Temperature:Ambient
CAS Number:112545-98-9
Formula:C₁₁H₁₂F₂O₃
MDL Number:MFCD11007701