Ethyl 2-(3,4-Difluorophenyl)-2,2-difluoroacetate
Catalog # APOSPC510340-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:236.163 g/mol
Storage Temperature:Ambient
CAS Number:73789-95-4
Formula:C₁₀H₈F₄O₂
MDL Number:MFCD11007709