Order Entry
Export
ContactUsLinkComponent
Ethyl 2,2-Difluoro-2-(p-tolyl)acetate
Ethyl 2,2-Difluoro-2-(p-tolyl)acetate
Catalog # APOSPC510337-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 2,2-Difluoro-2-(p-tolyl)acetate
Catalog # APOSPC510337-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :214.209 g/mol
Storage Temperature:Ambient
CAS Number:131323-06-3
Formula:C₁₁H₁₂F₂O₂
MDL Number:MFCD11007729