Ethyl 2-(2-Cyanophenyl)-2,2-difluoroacetate
Catalog # APOSPC510331-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:225.191 g/mol
Storage Temperature:Ambient
CAS Number:1335290-31-7
Formula:C₁₁H₉F₂NO₂
MDL Number:MFCD29065895