2-(2-Cyanophenyl)-2,2-difluoroacetic acid
Catalog # APOSPC510330-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:197.138 g/mol
Storage Temperature:Ambient
CAS Number:1824201-60-6
Formula:C₉H₅F₂NO₂
MDL Number:MFCD24502129