Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 2-(3-Bromophenyl)-2,2-difluoroacetate
Ethyl 2-(3-Bromophenyl)-2,2-difluoroacetate
Ethyl 2-(3-Bromophenyl)-2,2-difluoroacetate
Ethyl 2-(3-Bromophenyl)-2,2-difluoroacetate
Supplier:
Apollo Scientific
Certificates
Expand image
Ethyl 2-(3-Bromophenyl)-2,2-difluoroacetate
Supplier:
Apollo Scientific
About this item
CAS Number:
885068-75-7
:
Formula:
C₁₀H₉BrF₂O₂
MDL Number:
MFCD16036663
:
279.078 g/mol
Storage Temperature:
Ambient