Ethyl 2,2-Difluoro-2-(4-fluoro-3-methylphenyl)acetate
Catalog # APOSPC510323-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.199 g/mol
Storage Temperature:Ambient
CAS Number:1227160-98-6
Formula:C₁₁H₁₁F₃O₂
MDL Number:MFCD16036832