Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(4-Cyanophenyl)-2,2-difluoroacetic acid
2-(4-Cyanophenyl)-2,2-difluoroacetic acid
2-(4-Cyanophenyl)-2,2-difluoroacetic acid
Irritant
2-(4-Cyanophenyl)-2,2-difluoroacetic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-(4-Cyanophenyl)-2,2-difluoroacetic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
1261358-84-2
:
Formula:
C₉H₅F₂NO₂
MDL Number:
MFCD18533660
:
197.138 g/mol
:
GHS07
Storage Temperature:
Ambient