2,2-Difluoro-2-(2-nitrophenyl)acetic acid
Catalog # APOSPC510314-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:217.126 g/mol
Storage Temperature:Ambient
CAS Number:1308915-13-0
Formula:C₈H₅F₂NO₄
MDL Number:MFCD29065899