Ethyl 2,2-Difluoro-2-(1-naphthyl)acetate
Catalog # APOSPC510307-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:250.241 g/mol
Storage Temperature:Ambient
CAS Number:73790-00-8
Formula:C₁₄H₁₂F₂O₂
MDL Number:MFCD17064708