Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Dibromo-5-fluoro-1-methyl-1H-indazole
3,6-Dibromo-5-fluoro-1-methyl-1H-indazole
3,6-Dibromo-5-fluoro-1-methyl-1H-indazole
:
Apollo Scientific
3,6-Dibromo-5-fluoro-1-methyl-1H-indazole
:
Apollo Scientific
:
1286734-91-5
:
:
C₈H₅Br₂FN₂
:
MFCD18384875
:
307.945 g/mol
:
GHS07
:
Ambient