Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Dibromo-5-fluoro-1H-indazole
3,6-Dibromo-5-fluoro-1H-indazole
3,6-Dibromo-5-fluoro-1H-indazole
Irritant
3,6-Dibromo-5-fluoro-1H-indazole
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,6-Dibromo-5-fluoro-1H-indazole
Supplier:
Apollo Scientific
About this item
CAS Number:
1286734-81-3
:
Formula:
C₇H₃Br₂FN₂
MDL Number:
MFCD18384872
:
293.919 g/mol
:
GHS07
Storage Temperature:
Ambient