Ethyl 2,2-Difluoro-3-(methylamino)propanoate hydrochloride
Catalog # APOSPC510175-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:203.615 g/mol
Storage Temperature:Ambient
CAS Number:1956311-12-8
Formula:C₆H₁₁F₂NO₂·ClH
MDL Number:MFCD27986811