Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Difluoro-2-phenylethanamine
2,2-Difluoro-2-phenylethanamine
2,2-Difluoro-2-phenylethanamine
2,2-Difluoro-2-phenylethanamine
APOSPC510155-5G
:
Apollo Scientific
:
2,2-Difluoro-2-phenylethanamine
APOSPC510155-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
157.161 g/mol
:
Ambient
:
55601-21-3
:
C₈H₉F₂N
:
MFCD11100799