2-(1,1-Difluoroethyl)benzylamine
Catalog # APOSPC510154-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:171.187 g/mol
Storage Temperature:Ambient
CAS Number:1638767-86-8
Formula:C₉H₁₁F₂N
MDL Number:MFCD27500639