6-(Allyloxy)-2,3-difluorobenzyl Alcohol
Catalog # APOSPC510150-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:200.182 g/mol
Storage Temperature:Ambient
CAS Number:1823942-84-2
Formula:C₁₀H₁₀F₂O₂
MDL Number:MFCD26937565