

(2,4-Difluorophenyl)trimethylsilane
Catalog # APOSPC510134-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:186.274 g/mol
Boiling Pt:166…167 °C (747 mm Hg)
Density:1.049 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:148854-10-8
Formula:C₉H₁₂F₂Si
MDL Number:MFCD22682801