
2,2-Difluoro-3-(methylamino)propanoic acid hydrochloride
Catalog # APOSPC510102-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:175.562 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1346597-54-3
Formula:C₄H₇F₂NO₂·ClH
MDL Number:MFCD20491408