Order Entry
Export
ContactUsLinkComponent
3-(4-Bromo-2-fluorophenyl)-1,4-diazaspiro[4.4]nonan-2-one
3-(4-Bromo-2-fluorophenyl)-1,4-diazaspiro[4.4]nonan-2-one
Catalog # APOSPC510032-5G
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Bromo-2-fluorophenyl)-1,4-diazaspiro[4.4]nonan-2-one
Catalog # APOSPC510032-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :313.165 g/mol
Storage Temperature:Ambient
CAS Number:1272755-90-4
Formula:C₁₃H₁₄BrFN₂O
MDL Number:MFCD18651736