Order Entry
Export
ContactUsLinkComponent
5-Fluorospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-4-one
5-Fluorospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-4-one
Catalog # APOSPC510029-5G
Supplier:  Apollo Scientific
CAS Number:  
5-Fluorospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-4-one
Catalog # APOSPC510029-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :206.216 g/mol
Storage Temperature:Ambient
CAS Number:1272756-02-1
Formula:C₁₁H₁₁FN₂O
MDL Number:MFCD18651727