Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Benzyl 2-Fluoro-4-morpholinobenzoate
Benzyl 2-Fluoro-4-morpholinobenzoate
Benzyl 2-Fluoro-4-morpholinobenzoate
Benzyl 2-Fluoro-4-morpholinobenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Benzyl 2-Fluoro-4-morpholinobenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
1272756-24-7
:
Formula:
C₁₈H₁₈FNO₃
MDL Number:
MFCD18651702
:
315.339 g/mol
Storage Temperature:
Ambient