Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(2,6-Difluoro-4-nitrophenyl)propanoic acid
2-(2,6-Difluoro-4-nitrophenyl)propanoic acid
2-(2,6-Difluoro-4-nitrophenyl)propanoic acid
2-(2,6-Difluoro-4-nitrophenyl)propanoic acid
Supplier:
Apollo Scientific
Certificates
Expand image
2-(2,6-Difluoro-4-nitrophenyl)propanoic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
1226776-82-4
:
Formula:
C₉H₇F₂NO₄
MDL Number:
MFCD16495905
:
231.153 g/mol
Storage Temperature:
Ambient